accessibility menu, dialog, popup

UserName

(S)OHNOHHOOCH3

CAS RN 537-55-3

IUPAC Name: 2-acetamido-3-(4-hydroxyphenyl)propanoic acid
Synonyms: N-acetyl-L-tyrosine
Molecular Weight (g/mol): 223.23
Molecular Formula: C11H13NO4
InChi Key: CAHKINHBCWCHCF-UHFFFAOYNA-N
SMILES: CC(=O)NC(CC1=CC=C(O)C=C1)C(O)=O
Industry Type 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results