accessibility menu, dialog, popup

UserName

CH3ONHH3C

CAS RN 537-92-8

IUPAC Name: N-(3-methylphenyl)acetamide
Synonyms: 3-methylacetanilide
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: ALMHSXDYCFOZQD-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC(C)=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results