accessibility menu, dialog, popup

UserName

CAS RN 53902-12-8

H3COOHNOHOOCH3(E)

CAS RN 53902-12-8

IUPAC Name: 2-[(2E)-3-(3,4-dimethoxyphenyl)prop-2-enamido]benzoic acid
Synonyms: tranilast
Molecular Weight (g/mol): 327.34
Molecular Formula: C18H17NO5
InChi Key: NZHGWWWHIYHZNX-CSKARUKUSA-N
SMILES: COC1=CC=C(\C=C\C(=O)NC2=CC=CC=C2C(O)=O)C=C1OC
Physical Form 
  • (2)
  • (2)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)