accessibility menu, dialog, popup

UserName

CH3OOH3CCH3O

CAS RN 5394-63-8

IUPAC Name: 2,2,6-trimethyl-2,4-dihydro-1,3-dioxin-4-one
Synonyms: 2,2,6-trimethyl-1,3-dioxin-4-one
Molecular Weight (g/mol): 142.15
Molecular Formula: C7H10O3
InChi Key: XFRBXZCBOYNMJP-UHFFFAOYSA-N
SMILES: CC1=CC(=O)OC(C)(C)O1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)