missing translation for 'onlineSavingsMsg'
Learn More

2,2,6-Trimethyl-4H-1,3-dioxin-4-one, TRC

Product Code. 30451730
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451730 100 g 100g
30449344 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30451730 Supplier Toronto Research Chemicals Supplier No. TRCT796065100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5394-63-8
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.15
Synonym Diketene Acetone Adduct
IUPAC Name 2,2,6-trimethyl-1,3-dioxin-4-one
SMILES CC1=CC(=O)OC(C)(C)O1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C7H10O3/c1-5-4-6(8)10-7(2,3)9-5/h4H,1-3H3
Formula Weight 142.063
Chemical Name or Material 2,2,6-Trimethyl-4H-1,3-dioxin-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.