missing translation for 'onlineSavingsMsg'
Learn More

2,2,6-Trimethyl-4H-1,3-dioxin-4-one, 94%

Product Code. 10532841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10532841 100 g Glass bottle 100g
10107983 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10532841 Supplier Thermo Scientific Acros Supplier No. 296911000
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 5394-63-8 | C7H10O3 | 142.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5394-63-8
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.15
MDL Number MFCD00040468
InChI Key XFRBXZCBOYNMJP-UHFFFAOYSA-N
Synonym 2,2,6-trimethyl-4h-1,3-dioxin-4-one, 2,2,4-trimethyl-6-keto-1,3-dioxin, diketene acetone adduct, 4h-1,3-dioxin-4-one, 2,2,6-trimethyl, 2,2,6-trimethyl-1,3-dioxen-4-one, 2,2,6-trimethyl-1,3-dioxine-4-one, 2,2,6-trimethyl-1,3 dioxin-4-one, pubchem20364, 2,2,6-trimethyl-4h-1,3-dioxin-4-on, acmc-1akcb
PubChem CID 79368
IUPAC Name 2,2,6-trimethyl-1,3-dioxin-4-one
SMILES CC1=CC(=O)OC(O1)(C)C

Specifications

CAS Min % 90.0
CAS Max % 100.0
Melting Point 4°C to 13°C
Color Brown
Density 1.0800g/mL
Boiling Point 65°C to 67°C (2.7 mbar)
Flash Point 14°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Refractive Index 1.4590 to 1.462
Quantity 100 g
Beilstein 19, IV, 1604
Specific Gravity 1.08
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone and methanol
Formula Weight 142.15
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material 2,2,6-Trimethyl-4H-1,3-dioxin-4-one
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.