accessibility menu, dialog, popup

UserName

SHHS

CAS RN 540-63-6

IUPAC Name: ethane-1,2-dithiol
Synonyms: 1,2-ethanedithiol
Molecular Weight (g/mol): 94.19
Molecular Formula: C2H6S2
InChi Key: VYMPLPIFKRHAAC-UHFFFAOYSA-N
SMILES: SCCS
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)