missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Standard for ASTM D5623: Multi-component standard containing 22 sulfur species @ 50 μg/g (as Component) in Base Fuel

Product Code. 30310878
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30310878 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310878 Supplier VHG Labs Supplier No. VHGMSX222

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Ethanedithiol∣1,4-Butanedithiol∣1,5-Pentanedithiol∣1-Benzothiophene∣1-Decanethiol∣1-Heptanethiol∣1-Hexanethiol∣1-Nonanethiol∣1-Octanethiol∣1-Pentanethiol∣1-Propanethiol∣2-Ethylthiophene∣2-Methyl-1-propanethiol∣2-Methylthiophene∣3-Methylthiophene∣Carbon disulfide∣Dimethyl sulfide∣Methyl ethyl sulfide∣Propyl Disulfide∣Propyl Sulfide∣tert-Butyl disulfide∣tert-Butyl sulfide
CAS 540-63-6, 1191-08-8, 928-98-3, 95-15-8, 143-10-2, 1639-09-4
Analyte or Component Names 1,2-Ethanedithiol∣1,4-Butanedithiol∣1,5-Pentanedithiol∣1-Benzothiophene∣1-Decanethiol∣1-Heptanethiol∣1-Hexanethiol∣1-Nonanethiol∣1-Octanethiol∣1-Pentanethiol∣1-Propanethiol∣2-Ethylthiophene∣2-Methyl-1-propanethiol∣2-Methylthiophene∣3-Methylthiophene∣Carbon disulfide∣Dimethyl sulfide∣Methyl ethyl sulfide∣Propyl Disulfide∣Propyl Sulfide∣tert-Butyl disulfide∣tert-Butyl sulfide
Shipping Condition Room Temperature
Solution Type Single Solution
Solvent or Matrix Base Fuel
Industry Type Petrochemical Testing
Quantity 2 mL
Concentration or Composition (by Analyte or Components) 2-Methyl-1-propanethiol 50 μg/g∣1-Nonanethiol 50.1 μg/g∣2-Ethylthiophene 50.1 μg/g∣1,4-Butanedithiol 50.2 μg/g∣1-Decanethiol 50.2 μg/g∣tert-Butyl disulfide 50.2 μg/g∣Dimethyl sulfide 50.3 μg/g∣1,5-Pentanedithiol 50.4 μg/g∣1-Heptanethiol 50.4 μg/g∣Methyl ethyl sulfide 50.4 μg/g∣Propyl Disulfide 50.4 μg/g∣tert-Butyl sulfide 50.4 μg/g∣1-Pentanethiol 50.5 μg/g∣3-Methylthiophene 50.6 μg/g∣Propyl Sulfide 50.7 μg/g∣1-Octanethiol 50.9 μg/g∣1,2-Ethanedithiol 51 μg/g∣2-Methylthiophene 51 μg/g∣Carbon disulfide 51.5 μg/g∣1-Hexanethiol 51.8 μg/g∣1-Propanethiol 52.1 μg/g∣1-Benzothiophene 64.3 μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.