accessibility menu, dialog, popup

UserName

CH3HONOO

CAS RN 5428-54-6

IUPAC Name: 2-methyl-5-nitrophenol
Synonyms: 5-nitro-2-cresol
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: UMFDLIXUUJMPSI-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1O)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results