missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitrophenol, 97%, Thermo Scientific™

Product Code. 11424457
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424457 25 g 25g
11414457 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424457 Supplier Thermo Scientific Alfa Aesar Supplier No. B20472.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5428-54-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00043909
InChI Key UMFDLIXUUJMPSI-UHFFFAOYSA-N
Synonym 5-nitro-o-cresol, 5-nitro-2-cresol, phenol, 2-methyl-5-nitro, 2-hydroxy-4-nitrotoluene, o-cresol, 5-nitro, 5-nitro-2-methylphenol, 2-methy-5-nitrophenol, 2-methyl-5-nitro phenol, 2-methyl-5-nitro-phenol, 4-nitro-2-hydroxytoluene
PubChem CID 93576
IUPAC Name 2-methyl-5-nitrophenol
SMILES CC1=CC=C(C=C1O)[N+]([O-])=O

Specifications

Melting Point 116°C to 120°C
Boiling Point 180°C (15 mmHg)
Odor Phenol-like
Quantity 25 g
UN Number UN2446
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 2-Methyl-5-nitrophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.