accessibility menu, dialog, popup

UserName

NH2OHOHOO

CAS RN 5434-21-9

IUPAC Name: 4-aminobenzene-1,2-dicarboxylic acid
Synonyms: 4-aminobenzene-1,2-dicarboxylic acid
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: OXSANYRLJHSQEP-UHFFFAOYSA-N
SMILES: NC1=CC=C(C(O)=O)C(=C1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results