missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophthalic acid, 98%

Product Code. 11382106
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382106 5 g 5g
11372106 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382106 Supplier Thermo Scientific Alfa Aesar Supplier No. L03333.06
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5434-21-9 | C8H7NO4 | 181.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5434-21-9
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00013985
InChI Key OXSANYRLJHSQEP-UHFFFAOYSA-N
Synonym 4-aminobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-amino, 4-amino-1,2-benzenedicarboxylic acid, 4-amino phthalic acid, contains 3-aminophthalic acid, pubchem16853, acmc-1arqu, 4-aminophthalic acid, ksc270i4n, ghl.pd_mitscher_leg0.1208
PubChem CID 72912
IUPAC Name 4-aminophthalic acid
SMILES C1=CC(=C(C=C1N)C(=O)O)C(=O)O

Specifications

Melting Point >300°C
Odor Odorless
Quantity 5 g
Beilstein 2723405
Formula Weight 181.15
Percent Purity 98%
Chemical Name or Material 4-Aminophthalic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.