CAS RN 545-97-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Gibberellin A1, TRC
CAS: 545-97-1 Molecular Formula: C19 H24 O6 Molecular Weight (g/mol): 348.39 Synonym: 4aα,4bβ-Gibbane-1α,10β-dicarboxylic acid, 2β,4a,7-trihydroxy-1-methyl-8-methylene-, 1,4a-lactone (8CI),4a,1-(Epoxymethano)-7,9a-methanobenz[a]azulene, gibbane-1,10-dicarboxylic acid deriv.,(+)-Gibberillin A1,GA1,Giberellin A1 SMILES: C[C@]12[C@@H](O)CC[C@@]3(OC1=O)[C@@H]4CC[C@]5(O)C[C@]4(CC5=C)[C@H]([C@H]23)C(=O)O
2
Gibberellin A1 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality