missing translation for 'onlineSavingsMsg'
Learn More

Gibberellin A1, TRC

Product Code. 30006114
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006114 25 mg 25mg
17923265 5 mg 5mg
17991565 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006114 Supplier Toronto Research Chemicals Supplier No. TRCG37749525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 545-97-1
Molecular Formula C19 H24 O6
Molecular Weight (g/mol) 348.39
Synonym 4aα,4bβ-Gibbane-1α,10β-dicarboxylic acid, 2β,4a,7-trihydroxy-1-methyl-8-methylene-, 1,4a-lactone (8CI), 4a,1-(Epoxymethano)-7,9a-methanobenz[a]azulene, gibbane-1,10-dicarboxylic acid deriv., (+)-Gibberillin A1, GA1, Giberellin A1
SMILES C[C@]12[C@@H](O)CC[C@@]3(OC1=O)[C@@H]4CC[C@]5(O)C[C@]4(CC5=C)[C@H]([C@H]23)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H24O6/c1-9-7-17-8-18(9,24)5-3-10(17)19-6-4-11(20)16(2,15(23)25-19)13(19)12(17)14(21)22/h10-13,20,24H,1,3-8H2,2H3,(H,21,22)/t10-,11+,12-,13-,16-,17+,18+,19-/m1/s1
Formula Weight 348.1573
Percent Purity >95%
Chemical Name or Material Gibberellin A1
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.