CAS RN 54827-18-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress TMB monosulfate
MedChemExpress TMB monosulfate is a chromogenic substrate used in staining procedures in immunohistochemistry as well as being a visualizing reagent used in enzyme-linked immunosorbent assays (ELISA).