CAS RN 54903-09-2
IUPAC Name:
6-acetyl-2,3-dihydro-1,3-benzoxazol-2-one
Synonyms:
6-acetyl-3H-1,3-benzoxazol-2-one
Molecular Weight (g/mol):
177.16
Molecular Formula:
C9H7NO3
InChi Key:
QXBNAXVXLAHDTE-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=C2NC(=O)OC2=C1
1
–
1
of
1
results
1
6-Acetyl-2(3H)-benzoxazolone, 97%, Thermo Scientific Chemicals
CAS: 54903-09-2 Molecular Formula: C9H7NO3 Molecular Weight (g/mol): 177.159 MDL Number: MFCD01664312 InChI Key: QXBNAXVXLAHDTE-UHFFFAOYSA-N Synonym: 6-acetyl-2 3h-benzoxazolone,2 3h-benzoxazolone, 6-acetyl,jpb 2,6-acetyl-2-benzoxazolinone,6-acetylbenzo d oxazol-2 3h-one,6-acetyl-2,3-dihydro-1,3-benzoxazol-2-one,acetyl-6 benzoxazolinone french,6-acetyl-3h-benzooxazol-2-one,6-acetyl-1,3-benzoxazol-2 3h-one,6-acetyl-2-oxo-3-hydrobenzoxazole PubChem CID: 689054 IUPAC Name: 6-acetyl-3H-1,3-benzoxazol-2-one SMILES: CC(=O)C1=CC2=C(C=C1)NC(=O)O2