missing translation for 'onlineSavingsMsg'
Learn More

6-Acetyl-2(3H)-benzoxazolone, 97%, Thermo Scientific Chemicals

Product Code. 11307149
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11307149 5 g 5g
11317149 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307149 Supplier Thermo Scientific Alfa Aesar Supplier No. H27368.06
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 54903-09-2 | Molecular Formula: C9H7NO3 | Molecular Weight: 177.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54903-09-2
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.159
MDL Number MFCD01664312
InChI Key QXBNAXVXLAHDTE-UHFFFAOYSA-N
Synonym 6-acetyl-2 3h-benzoxazolone, 2 3h-benzoxazolone, 6-acetyl, jpb 2, 6-acetyl-2-benzoxazolinone, 6-acetylbenzo d oxazol-2 3h-one, 6-acetyl-2,3-dihydro-1,3-benzoxazol-2-one, acetyl-6 benzoxazolinone french, 6-acetyl-3h-benzooxazol-2-one, 6-acetyl-1,3-benzoxazol-2 3h-one, 6-acetyl-2-oxo-3-hydrobenzoxazole
PubChem CID 689054
IUPAC Name 6-acetyl-3H-1,3-benzoxazol-2-one
SMILES CC(=O)C1=CC2=C(C=C1)NC(=O)O2

Specifications

Melting Point 232°C to 239°C
Quantity 5 g
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material 6-Acetyl-2(3H)-benzoxazolone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.