accessibility menu, dialog, popup

UserName

HNSOONOO

CAS RN 549476-61-1

IUPAC Name: N-cyclopropyl-4-nitrobenzene-1-sulfonamide
Synonyms: N-cyclopropyl-4-nitrobenzenesulfonamide
Molecular Weight (g/mol): 242.25
Molecular Formula: C9H10N2O4S
InChi Key: SFXOKDPLGLHGIX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)S(=O)(=O)NC1CC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results