missing translation for 'onlineSavingsMsg'
Learn More

N-Cyclopropyl-4-nitrobenzenesulfonamide, 97%, Thermo Scientific™

Product Code. 15424908
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424908 500 mg 500mg
15414908 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424908 Supplier Thermo Scientific Alfa Aesar Supplier No. H58673.ME
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 549476-61-1
Molecular Formula C9H10N2O4S
Molecular Weight (g/mol) 242.249
MDL Number MFCD00458256
InChI Key SFXOKDPLGLHGIX-UHFFFAOYSA-N
PubChem CID 3434716
IUPAC Name N-cyclopropyl-4-nitrobenzenesulfonamide
SMILES C1CC1NS(=O)(=O)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Quantity 500 mg
Synonym n-cyclopropyl 4-nitrophenylsulfonamide, cyclopropyl 4-nitrophenyl sulfonyl amine, n-cyclopropyl4-nitrophenylsulfonamide, pubchem18638, acmc-209lkb, n-cyclopropyl-4-nitro-benzenesulfonamide, n-cyclopropyl-4-nitrobenzene-1-sulfonamide
Formula Weight 242.25
Percent Purity 97%
Chemical Name or Material N-Cyclopropyl-4-nitrobenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.