accessibility menu, dialog, popup

UserName

CAS RN 55117-15-2

IUPAC Name: 1-chloro-2-(chloromethyl)-3-fluorobenzene
Synonyms: 1-chloro-2-(chloromethyl)-3-fluorobenzene
Molecular Weight (g/mol): 179.02
Molecular Formula: C7H5Cl2F
InChi Key: MJGOLNNLNQQIHR-UHFFFAOYSA-N
SMILES: FC1=CC=CC(Cl)=C1CCl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results