accessibility menu, dialog, popup

UserName

CAS RN 5535-49-9

IUPAC Name: [(2-chloroethyl)sulfanyl]benzene
Synonyms: [(2-chloroethyl)sulfanyl]benzene
Molecular Weight (g/mol): 172.67
Molecular Formula: C8H9ClS
InChi Key: QDXIHHOPZFCEAP-UHFFFAOYSA-N
SMILES: ClCCSC1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results