missing translation for 'onlineSavingsMsg'
Learn More

2-chloroethylephenyl sulfide, 98%, Thermo Scientific™

Product Code. 11376410 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5mL; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11376410 5mL; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11376410 Supplier Thermo Scientific Acros Supplier No. 302210050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5535-49-9
CAS Min % 97.5
CAS Max % 100.0
Density 1.1700g/mL
Boiling Point 245.0°C to 247.0°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H9ClS
Refractive Index 1.5820 to 1.5840
MDL Number MFCD00013691
Quantity 5mL
Specific Gravity 1.17
Synonym 2-chloroethyl phenyl sulfide, 2-chloroethyl phenyl sulfane, 2-chloroethyl sulfanyl benzene, 2-chloroethyl thio benzene, 2-chloroethylphenylsulfide, benzene, 2-chloroethyl thio, 2-phenylthio ethyl chloride, 2-phenylthioethylchloride, acmc-1axe6
InChI Key QDXIHHOPZFCEAP-UHFFFAOYSA-N
SMILES ClCCSC1=CC=CC=C1
IUPAC Name [(2-chloroethyl)sulfanyl]benzene
Molecular Weight (g/mol) 172.67
PubChem CID 21702
Formula Weight 172.68
Percent Purity 98%
Chemical Name or Material 2-Chloroethyl phenyl sulfide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.