accessibility menu, dialog, popup

UserName

CAS RN 554-01-8

CH3NH2NHON

CAS RN 554-01-8

IUPAC Name: 6-amino-5-methyl-1,2-dihydropyrimidin-2-one
Synonyms: 5-methylcytosine
Molecular Weight (g/mol): 125.13
Molecular Formula: C5H7N3O
InChi Key: LRSASMSXMSNRBT-UHFFFAOYSA-N
SMILES: CC1=C(N)NC(=O)N=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)