accessibility menu, dialog, popup

UserName

OHOHOCl

CAS RN 56363-84-9

IUPAC Name: 2-chloro-4-hydroxybenzoic acid
Synonyms: 2-chloro-4-hydroxybenzoic acid
Molecular Weight (g/mol): 172.56
Molecular Formula: C7H5ClO3
InChi Key: WIPYZRZPNMUSER-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(O)C=C1Cl
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results