missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-4-hydroxybenzoic acid hydrate, 98%

Product Code. 10481301 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10481301 1 g Glass bottle 1g
10388993 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10481301 Supplier Thermo Scientific Acros Supplier No. 304200010
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56363-84-9 | C7H5ClO3 | 172.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56363-84-9
Molecular Formula C7H5ClO3
Molecular Weight (g/mol) 172.56
MDL Number MFCD00798116
InChI Key WIPYZRZPNMUSER-UHFFFAOYSA-N
Synonym benzoic acid, 2-chloro-4-hydroxy, 2-chloro-4-hydroxy-benzoic acid, 2-chloro-4-hyroxybenzoic acid, pubchem4561, acmc-1b14n, ksc497o6l, 4-hydroxy-2-chlorobenzoic acid, wipyzrzpnmuser-uhfffaoysa
PubChem CID 91821
IUPAC Name 2-chloro-4-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(O)C=C1Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 207°C to 209°C
Color Beige to White or Brown-Orange
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 172.57
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloro-4-hydroxybenzoic acid hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.