CAS RN 566-75-6
CAS RN 566-75-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
16-Keto 17Beta-Estradiol, TRC
CAS: 566-75-6 Molecular Formula: C18H22O3 Molecular Weight (g/mol): 286.37 Synonym: 16-Oxoestradiol IUPAC Name: (8R,9S,13S,14S,17R)-3,17-dihydroxy-13-methyl-7,8,9,11,12,14,15,17-octahydro-6H-cyclopenta[a]phenanthren-16-one SMILES: C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC(=O)[C@@H]2O
2
16-Oxoestradiol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.