accessibility menu, dialog, popup

UserName

H3COBOHHO

CAS RN 5720-06-9

IUPAC Name: (2-methoxyphenyl)boronic acid
Synonyms: 2-methoxyphenylboronic acid
Molecular Weight (g/mol): 151.96
Molecular Formula: C7H9BO3
InChi Key: ROEQGIFOWRQYHD-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1B(O)O
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results