missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenylboronic acid, 97%

Product Code. 10786442 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10786442 5 g Glass bottle 5g
10603722 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10786442 Supplier Thermo Scientific Acros Supplier No. 344660050
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 5720-06-9 | C7H9BO3 | 151.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 5720-06-9
Molecular Formula C7H9BO3
Molecular Weight (g/mol) 151.96
MDL Number MFCD00236047
InChI Key ROEQGIFOWRQYHD-UHFFFAOYSA-N
SMILES COC1=CC=CC=C1B(O)O

Spezifikation

CAS Min % 96.0
CAS Max % 100.0
Melting Point 106.0°C to 111.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3OC6H4B(OH)2
Quantity 5 g
Synonym 4-acetyl-4'-bromobiphenyl, 4'-4-bromophenyl acetophenone, 1-4-4-bromophenyl phenyl ethanone, 4'-p-bromophenyl acetophenone, 1-4'-bromo-1,1'-biphenyl-4-yl ethanone, 4-acetyl-4'-bromodiphenyl, 4-bromo-4'-acetylbiphenyl, ethanone, 1-4'-bromo 1,1'-biphenyl-4-yl, 1-4'-bromo 1,1'-biphenyl-4-yl ethanone, 1-4'-bromo 1,1'-biphenyl-4-yl ethan-1-one
Solubility Information Solubility in water:+/- 2.5% in water
IUPAC Name 1-[4-(4-bromophenyl)phenyl]ethanone
PubChem CID 79806
Formula Weight 151.96
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Methoxyphenylboronic acid, 97%
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 2-Methoxyphenylboronic acid
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.