accessibility menu, dialog, popup

UserName

CAS RN 57568-60-2

IUPAC Name: (2E)-3-(furan-2-yl)-2-phenylprop-2-enal
Synonyms: (2E)-3-(furan-2-yl)-2-phenylprop-2-enal
Molecular Weight (g/mol): 198.22
Molecular Formula: C13H10O2
InChi Key: JPESOGFYFXAURP-XFXZXTDPSA-N
SMILES: O=C\C(=C\C1=CC=CO1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results