missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)-2-phenylpropenal, 97%

Product Code. 11312426
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312426 25 g 25g
11302426 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312426 Supplier Thermo Scientific Alfa Aesar Supplier No. L06220.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57568-60-2 | C13H10O2 | 198.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 57568-60-2
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00036566
InChI Key JPESOGFYFXAURP-XFXZXTDPSA-N
Synonym 3-2-furyl-2-phenylpropenal, 3-2'-furyl-2-phenyl-2-propenal, 3-2-furyl-2-phenyl propenal, z-3-furan-2-yl-2-phenylprop-2-enal, 2z-3-furan-2-yl-2-phenylprop-2-enal
PubChem CID 6537126
IUPAC Name (Z)-3-(furan-2-yl)-2-phenylprop-2-enal
SMILES O=C\C(=C\C1=CC=CO1)C1=CC=CC=C1

Especificaciones

Melting Point 57°C to 60°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 5923
Sensitivity Air Sensitive
Formula Weight 198.22
Percent Purity 97%
Chemical Name or Material 3-(2-Furyl)-2-phenylpropenal

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.