accessibility menu, dialog, popup

UserName

BrSOOCl

CAS RN 58236-74-1

IUPAC Name: (3-bromophenyl)methanesulfonyl chloride
Synonyms: (3-bromophenyl)methanesulfonyl chloride
Molecular Weight (g/mol): 269.54
Molecular Formula: C7H6BrClO2S
InChi Key: VCGKEAIINMDLMC-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)CC1=CC(Br)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results