missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzylsulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10703874
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10703874 1 g 1g
10185483 5 g 5g
10784087 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10703874 Supplier Thermo Scientific Maybridge Supplier No. MO07946DA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 58236-74-1
Molecular Formula C7H6BrClO2S
Molecular Weight (g/mol) 269.537
MDL Number MFCD06798043
InChI Key VCGKEAIINMDLMC-UHFFFAOYSA-N
Synonym 3-bromophenyl methanesulfonyl chloride, 3-bromobenzylsulfonyl chloride, 3-bromophenyl methanesulphonyl chloride, benzenemethanesulfonylchloride, 3-bromo, benzenemethanesulfonyl chloride, 3-bromo, 3-bromobenzylsulphonyl chloride, 3-bromophenyl methylsulphonyl chloride
PubChem CID 2761451
IUPAC Name (3-bromophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC(=C1)Br)CS(=O)(=O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 269.55
Percent Purity 97%
Chemical Name or Material 3-Bromobenzylsulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.