accessibility menu, dialog, popup

UserName

H3CCH3(R)CH3(R)CH3(R)H3CCH3OHH3CH3CO

CAS RN 59-02-9

IUPAC Name: (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
Synonyms: vitamin E snowe muscle, energy & feritility bayer, tocopherol E mulsin togasan vitamin e togasan vitamin E tocopherol rodisma-med brand E, auxina e hydrosol, vit E, puncto
Molecular Weight (g/mol): 430.72
Molecular Formula: C29H50O2
InChi Key: GVJHHUAWPYXKBD-IEOSBIPESA-N
SMILES: CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results