accessibility menu, dialog, popup

UserName

CH3SnH3CCH3N

CAS RN 59020-10-9

IUPAC Name: 3-(tributylstannyl)pyridine
Synonyms: 3-(tributylstannyl)pyridine
Molecular Weight (g/mol): 368.15
Molecular Formula: C17H31NSn
InChi Key: CFQJBWKKHCMCGJ-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=CN=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)