missing translation for 'onlineSavingsMsg'
Learn More

3-(1,1,1-Tributylstannyl)pyridine, 90%, Thermo Scientific™

Product Code. 10492274
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10492274 5 g Amber Glass Bottle 5g
10437052 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10492274 Supplier Thermo Scientific Maybridge Supplier No. KM05818DE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 59020-10-9
Molecular Formula C17H31NSn
Molecular Weight (g/mol) 368.152
MDL Number MFCD00052054
InChI Key CFQJBWKKHCMCGJ-UHFFFAOYSA-N
Synonym 3-tributylstannyl pyridine, 3-1,1,1-tributylstannyl pyridine, 3-pyridyltri-n-butyltin, pyridine, 3-tributylstannyl, 3-tri-n-butylstannyl pyridine, 3-pyridyltributylstannane, 3-tributylstannylpyridine, 3-tris but-1-ylstannyl pyridine, 3-tributylstannanyl-pyridine, 3-pyridyltributyltin
PubChem CID 3483848
IUPAC Name tributyl(pyridin-3-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CN=CC=C1

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 368.15
Percent Purity 90%
Chemical Name or Material 3-(1, 1, 1-Tributylstannyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.