accessibility menu, dialog, popup

UserName

H2ONaH3CNSOOOCH3NCH3NO

CAS RN 5907-38-0

IUPAC Name: sodium [(1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)(methyl)amino]methanesulfonate hydrate
Synonyms: sodium metamizole(1-) hydrate
Molecular Weight (g/mol): 351.35
Molecular Formula: C13H18N3NaO5S
InChi Key: UNZIDPIPYUMVPA-UHFFFAOYSA-M
SMILES: O.[Na+].CN(CS([O-])(=O)=O)C1=C(C)N(C)N(C1=O)C1=CC=CC=C1
Certifications/Compliance 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Industry Type 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)