accessibility menu, dialog, popup

UserName

CAS RN 593-51-1

ClCH3H3N

CAS RN 593-51-1

IUPAC Name: methanaminium chloride
Synonyms: methylamine hydrochloride monomethylamine hydrochloride methanamine, hydrochloride (1:1)
Molecular Weight (g/mol): 67.52
Molecular Formula: CH6ClN
InChi Key: NQMRYBIKMRVZLB-UHFFFAOYSA-N
SMILES: [Cl-].C[NH3+]
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (5)
  • (3)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)