accessibility menu, dialog, popup

UserName

OHOOHHOOH

CAS RN 60-82-2

IUPAC Name: 3-(4-hydroxyphenyl)-1-(2,4,6-trihydroxyphenyl)propan-1-one
Synonyms: phloretin 根皮素 dihydronaringenin 4-O-methylphloracetophenone phloretol
Molecular Weight (g/mol): 274.27
Molecular Formula: C15H14O5
InChi Key: VGEREEWJJVICBM-UHFFFAOYSA-N
SMILES: OC1=CC=C(CCC(=O)C2=C(O)C=C(O)C=C2O)C=C1
Molecular Weight (g/mol) 
  • (7)
  • (2)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (4)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (7)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)