accessibility menu, dialog, popup

UserName

H3COOHNNH2OCH3

CAS RN 60075-23-2

IUPAC Name: 2-(3,4-dimethoxyphenyl)acetohydrazide
Synonyms: 2-(3,4-dimethoxyphenyl)acetohydrazide
Molecular Weight (g/mol): 210.23
Molecular Formula: C10H14N2O3
InChi Key: HRMXYTRKEOUMNG-UHFFFAOYSA-N
SMILES: COC1=CC=C(CC(=O)NN)C=C1OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results