missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyphenylacetic acid hydrazide, 97%, Thermo Scientific™

Product Code. 11468893
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468893 25 g 25g
11448893 1 g 1g
11458893 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468893 Supplier Thermo Scientific Alfa Aesar Supplier No. A11049.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60075-23-2
Molecular Formula C10H14N2O3
Molecular Weight (g/mol) 210.23
MDL Number MFCD00051701
InChI Key HRMXYTRKEOUMNG-UHFFFAOYSA-N
Synonym 2-3,4-dimethoxyphenyl acetohydrazide, 3,4-dimethoxyphenylacetic acid hydrazide, 3,4-dimethoxyphenylacethydrazide, 3,4-dimethoxyphenyl acetic hydrazide, acmc-20aneo, homoveratric hydrazide, maybridge1_007491, 2-3,4-dimethoxyphenyl ethanehydrazide, 2-3,4-dimethoxyphenyl ethanohydrazide, 2-3,4-dimethoxyphenyl acetic hydrazide
PubChem CID 282482
IUPAC Name 2-(3,4-dimethoxyphenyl)acetohydrazide
SMILES COC1=CC=C(CC(=O)NN)C=C1OC

Specifications

Melting Point 102°C to 104°C
Quantity 25 g
Beilstein 2698478
Formula Weight 210.23
Percent Purity 97%
Chemical Name or Material 3,4-Dimethoxyphenylacetic acid hydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.