accessibility menu, dialog, popup

UserName

CH3OONOO

CAS RN 606-27-9

IUPAC Name: methyl 2-nitrobenzoate
Synonyms: methyl 2-nitrobenzoate
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: AOXPHVNMBPFOFS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1[N+]([O-])=O
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results