missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-nitrobenzoate, 98+%

Product Code. 11496176
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496176 100 g 100g
11486176 25 g 25g
11406186 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496176 Supplier Thermo Scientific Alfa Aesar Supplier No. B23009.22
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 606-27-9 | C8H7NO4 | 181.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-27-9
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007136
InChI Key AOXPHVNMBPFOFS-UHFFFAOYSA-N
Synonym methyl o-nitrobenzoate, benzoic acid, 2-nitro-, methyl ester, 2-nitro-benzoic acid methyl ester, benzoic acid, o-nitro-, methyl ester, 2-nitrobenzoic acid methyl ester, methyl-2-nitrobenzoate, methyl o-nitro benzoate, methyl ortho-nitrobenzoate, acmc-209ml4, nitrobenzoic acid methyl ester
PubChem CID 69072
IUPAC Name methyl 2-nitrobenzoate
SMILES COC(=O)C1=CC=CC=C1[N+](=O)[O-]

Specifications

Melting Point -13°C
Density 1.285
Boiling Point 275°C
Flash Point >110°C (230°F)
Refractive Index 1.533
Quantity 100 g
Beilstein 2050101
Formula Weight 181.15
Percent Purity ≥98%
Chemical Name or Material Methyl 2-nitrobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.