accessibility menu, dialog, popup

UserName

CAS RN 6080-58-6

IUPAC Name: trilithium(1+) 2-hydroxypropane-1,2,3-tricarboxylate tetrahydrate
Synonyms: trilithium(1+) citrate tetrahydrate
Molecular Weight (g/mol): 281.98
Molecular Formula: C6H13Li3O11
InChi Key: HXGWMCJZLNWEBC-UHFFFAOYSA-K
SMILES: [Li+].[Li+].[Li+].O.O.O.O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Grade 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)