missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, trilithium salt tetrahydrate, 98%, pure, Thermo Scientific Chemicals

Product Code. 10286730 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
500g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10286730 500 g Plastic bottle 500g
10511841 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10286730 Supplier Thermo Scientific Acros Supplier No. 199855000
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6080-58-6 | Molecular Formula: C6H5Li3O7·4H2O | Molecular Weight: 281.979 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6080-58-6
MDL Number MFCD00150030
InChI Key HXGWMCJZLNWEBC-UHFFFAOYSA-K
Synonym lithium citrate tetrahydrate, unii-5z6e9k79yv, trilithium citrate tetrahydrate, lithium citrate usp, citric acid trilithium salt tetrahydrate, tri-lithium citrate tetrahydrate, lithium citrate tribasic tetrahydrate, lithonate tn, 1,2,3-propanetricarboxylic acid, 2-hydroxy-trilithium salt tetrahydrate
PubChem CID 2724118
ChEBI CHEBI:64754
IUPAC Name trilithium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate
SMILES [Li+].[Li+].[Li+].C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O

Specifications

Melting Point 112.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Molecular Formula C6H5Li3O7·4H2O
Linear Formula LiO2CCH2C(OH)(CO2Li)CH2CO2Li·4H2O
Quantity 500 g
Beilstein 03, I, 196
Merck Index 15, 5586
Solubility Information Solubility in water: 745g/L water (25°C). Other solubilities: slightly soluble in alcohol
Molecular Weight (g/mol) 281.97
Formula Weight 281.97
Percent Purity 98%
Grade Pure
Physical Form Fine Crystalline Powder
Chemical Name or Material Citric acid, trilithium salt tetrahydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.