accessibility menu, dialog, popup

UserName

OHIII

CAS RN 609-23-4

IUPAC Name: 2,4,6-triiodophenol
Synonyms: 2,4,6-triiodophenol 246triiodophenol
Molecular Weight (g/mol): 471.8
Molecular Formula: C6H3I3O
InChi Key: VAPDZNUFNKUROY-UHFFFAOYSA-N
SMILES: OC1=C(I)C=C(I)C=C1I
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results