missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triiodophenol, 98%

Product Code. 11433367
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433367 100 g 100g
11414100 5 g 5g
11423367 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433367 Supplier Thermo Scientific Alfa Aesar Supplier No. A17145.22
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 609-23-4 | C6H3I3O | 471.802 g/mol

2,4,6-Triiodophenol is used as an iodinated disinfection by product formed during chlorination of sewage effluents. It is also used as a potent thyroid disrupting chemical and to investigate deiodinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Triiodophenol is used as an iodinated disinfection by product formed during chlorination of sewage effluents. It is also used as a potent thyroid disrupting chemical and to investigate deiodinase.

Solubility
Insoluble in water.

Notes
Incompatible withacid chlorides, acid anhydrides and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-23-4
Molecular Formula C6H3I3O
Molecular Weight (g/mol) 471.802
MDL Number MFCD00002179
InChI Key VAPDZNUFNKUROY-UHFFFAOYSA-N
Synonym phenol, 2,4,6-triiodo, triiodophenol, unii-9rb2r81a7u, 2,4,6-trijodfenol, 2,4,6-trijodfenol czech, 2,4,6-triiodo-phenol, chembl81721, 2,4,6-triiodo-phenol tip, 2,6-triiodophenol, phenol,4,6-triiodo
PubChem CID 11862
IUPAC Name 2,4,6-triiodophenol
SMILES C1=C(C=C(C(=C1I)O)I)I

Specifications

Melting Point 156°C to 160°C
Quantity 100 g
Beilstein 2046861
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 471.8
Percent Purity 98%
Chemical Name or Material 2,4,6-Triiodophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.