accessibility menu, dialog, popup

UserName

CAS RN 6091-64-1

H3COOBr

CAS RN 6091-64-1

IUPAC Name: ethyl 2-bromobenzoate
Synonyms: ethyl 2-bromobenzoate
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: BIHHBTVQFPVSTE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1Br
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)