missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromobenzoate, 98+%

Product Code. 11444554
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. Quantity unitSize
11444554 250 g 250g
11424554 10 g 10g
11434554 50 g 50g
3 options
This item is not returnable. View return policy

Product Code. 11444554

Brand: Thermo Scientific Alfa Aesar A13166.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6091-64-1 | C9H9BrO2 | 229.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6091-64-1
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00015443
InChI Key BIHHBTVQFPVSTE-UHFFFAOYSA-N
Synonym 2-bromobenzoic acid ethyl ester, benzoic acid, 2-bromo-, ethyl ester, ethyl-2-bromobenzoate, ethyl2-bromobenzoate, ethyl bromobenzoate, ethyl o-bromobenzoate, pubchem3937, brombenzoesaureathylester, acmc-209mn8, ksc493o4r
PubChem CID 80186
IUPAC Name ethyl 2-bromobenzoate
SMILES CCOC(=O)C1=CC=CC=C1Br

Specifications

Density 1.44
Boiling Point 255°C
Flash Point >110°C (230°F)
Refractive Index 1.544
Quantity 250 g
Beilstein 2207360
Formula Weight 229.07
Percent Purity ≥98%
Chemical Name or Material Ethyl 2-bromobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.