accessibility menu, dialog, popup

UserName

HOOO

CAS RN 6093-68-1

IUPAC Name: 6-hydroxy-2H-chromen-2-one
Synonyms: 6-hydroxychromen-2-one
Molecular Weight (g/mol): 162.14
Molecular Formula: C9H6O3
InChi Key: CJIJXIFQYOPWTF-UHFFFAOYSA-N
SMILES: OC1=CC=C2OC(=O)C=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results