missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxycoumarin, 98%

Product Code. 15590624 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15590624 1 g 1g
15500634 5 g 5g
15510634 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15590624 Supplier Thermo Scientific Acros Supplier No. 459410010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6093-68-1 | C9H6O3 | 162.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6093-68-1
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.15
InChI Key CJIJXIFQYOPWTF-UHFFFAOYSA-N
Synonym 6-hydroxycoumarin, 6-hydroxy-2h-chromen-2-one, 2h-1-benzopyran-2-one, 6-hydroxy, 6-hydroxycoumaran, 2h-1-benzopyran-2-one,6-hydroxy, 6-hydroxbenzopyran-2-one, 6-hydroxy-2h-chromen-2-on, 6-hydroxycoumarine, 6-hydroxy coumarin, 6-hydroxy-2h-1-benzopyran-2-one
PubChem CID 99477
IUPAC Name 6-hydroxychromen-2-one
SMILES C1=CC2=C(C=CC(=O)O2)C=C1O

Specifications

Melting Point 249.0°C to 253.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 162.15
Percent Purity 98%
Chemical Name or Material 6-Hydroxycoumarin
compliance-icons
Product Identifier
  • 6-Hydroxycoumarin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.